accessibility menu, dialog, popup

UserName

CAS RN 1150114-28-5

BFNOOFHOOH

CAS RN 1150114-28-5

IUPAC Name: (2,6-difluoro-3-nitrophenyl)boronic acid
Synonyms: 2,6-difluoro-3-nitrophenylboronic acid
Molecular Weight (g/mol): 202.91
Molecular Formula: C6H4BF2NO4
InChi Key: SPEDPBJIXAVPMJ-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC(=C1F)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results