accessibility menu, dialog, popup

UserName

CAS RN 113886-71-8

CH3ONHNONOOHOHOCH3

CAS RN 113886-71-8

IUPAC Name: N-{1-[4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}acetamide
Synonyms: N-{1-[4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-2-oxopyrimidin-4-yl}acetamide
Molecular Weight (g/mol): 299.28
Molecular Formula: C12H17N3O6
InChi Key: CYDFBLGNJUNSCC-UHFFFAOYNA-N
SMILES: COC1C(O)C(CO)OC1N1C=CC(NC(C)=O)=NC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)