accessibility menu, dialog, popup

UserName

CAS RN 112883-42-8

H3C(S)OHONCH3OO

CAS RN 112883-42-8

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)hexanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}hexanoic acid
Molecular Weight (g/mol): 367.45
Molecular Formula: C22H25NO4
InChi Key: RZZXDYZWHUAOEK-UHFFFAOYNA-N
SMILES: CCCCC(N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)