CAS RN 112641-20-0
CAS RN 112641-20-0
IUPAC Name:
2-fluoro-3-(trifluoromethyl)benzaldehyde
Synonyms:
2-fluoro-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol):
192.11
Molecular Formula:
C8H4F4O
InChi Key:
XDMZVNQKVMTCSP-UHFFFAOYSA-N
SMILES:
FC1=C(C=O)C=CC=C1C(F)(F)F
1
–
1
of
1
results
1
2-Fluoro-3-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
CAS: 112641-20-0 Molecular Formula: C8H4F4O Molecular Weight (g/mol): 192.113 MDL Number: MFCD00061246 InChI Key: XDMZVNQKVMTCSP-UHFFFAOYSA-N PubChem CID: 517978 IUPAC Name: 2-fluoro-3-(trifluoromethyl)benzaldehyde SMILES: C1=CC(=C(C(=C1)C(F)(F)F)F)C=O