accessibility menu, dialog, popup

UserName

CAS RN 1123-34-8

HOCH2

CAS RN 1123-34-8

IUPAC Name: 1-(prop-2-en-1-yl)cyclohexan-1-ol
Synonyms: 1-(prop-2-en-1-yl)cyclohexan-1-ol
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: ZSLKGUQYEQVKQE-UHFFFAOYSA-N
SMILES: OC1(CC=C)CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results