accessibility menu, dialog, popup

UserName

CAS RN 112193-41-6

NH2HNONBr

CAS RN 112193-41-6

IUPAC Name: 5-bromopyridine-3-carbohydrazide
Synonyms: 5-bromopyridine-3-carbohydrazide
Molecular Weight (g/mol): 216.04
Molecular Formula: C6H6BrN3O
InChi Key: VERXTAUONOFMLH-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CN=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results