accessibility menu, dialog, popup

UserName

CAS RN 110-78-1

CH3NCO

CAS RN 110-78-1

IUPAC Name: 1-isocyanatopropane
Synonyms: propane, 1-isocyanato-
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: OQURWGJAWSLGQG-UHFFFAOYSA-N
SMILES: CCCN=C=O
Boiling Point 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)