accessibility menu, dialog, popup

UserName

CAS RN 109838-85-9

H3CON(R)CH3CH3NOCH3

CAS RN 109838-85-9

IUPAC Name: (2R)-3,6-dimethoxy-2-(propan-2-yl)-2,5-dihydropyrazine
Synonyms: (3R)-3-isopropyl-2,5-dimethoxy-3,6-dihydropyrazine
Molecular Weight (g/mol): 184.24
Molecular Formula: C9H16N2O2
InChi Key: FCFWEOGTZZPCTO-MRVPVSSYSA-N
SMILES: COC1=N[C@H](C(C)C)C(OC)=NC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)