accessibility menu, dialog, popup

UserName

CAS RN 109-04-6

BrN

CAS RN 109-04-6

IUPAC Name: 2-bromopyridine
Synonyms: 2-bromopyridine
Molecular Weight (g/mol): 158
Molecular Formula: C5H4BrN
InChi Key: IMRWILPUOVGIMU-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=N1
Boiling Point 
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)