CAS RN 108928-81-0
CAS RN 108928-81-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxy-Alpha1-[[[6-(1-methyl-3-phenylpropoxy)hexyl]amino]methyl]-1,3-benzenedimethanol (Salmeterol Impurity), TRC
CAS: 108928-81-0 Molecular Formula: C25 H37 N O4 Molecular Weight (g/mol): 415.57 Synonym: 1,3-Benzenedimethanol, 4-hydroxy-α1-[[[6-(1-methyl-3-phenylpropoxy)hexyl]amino]methyl]-,4-{1-Hydroxy-2-[6-(4-phenylbutan-2-yloxy)hexylamino]ethyl}-2-(hydroxymethyl)phenol,1-[4-Hydroxy-3-(hydroxymethyl)-phenyl]-2-[[6-(1-methyl-3-phenylpropoxy)hexyl]-amino]ethanol,Salmeterol Related Compound B (USP),Salmeterol Xinafoate Imp. E (EP),GR 58583X,USP Salmeterol Related Compound B,Ph Eur Salmeterol Impurity E,GR 58583X IUPAC Name: 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutan-2-yloxy)hexylamino]ethyl]phenol SMILES: CC(CCc1ccccc1)OCCCCCCNCC(O)c2ccc(O)c(CO)c2