accessibility menu, dialog, popup

UserName

CAS RN 108750-13-6

OOONOOOOOHHONOHOOHONaNa

CAS RN 108750-13-6

IUPAC Name: disodium 2-{[(2'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-7'-yl)methyl](carboxylatomethyl)amino}acetate
Synonyms: disodium 2-[(2'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthen]-7'-ylmethyl)(carboxylatomethyl)amino]acetate
Molecular Weight (g/mol): 666.5
Molecular Formula: C30H24N2Na2O13
InChi Key: ZPDXOJYVKULICC-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OC(=O)CN(CC(O)=O)CC1=C(O)C=C2OC3=CC(O)=C(CN(CC([O-])=O)CC([O-])=O)C=C3C3(OC(=O)C4=CC=CC=C34)C2=C1
Color 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results