CAS RN 108374-32-9
CAS RN 108374-32-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
No Refinements
1
–
0
of
0
results
1
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
CAS: 108374-32-9 Molecular Formula: Cl2H12N4Pt Molecular Weight (g/mol): 334.11 MDL Number: MFCD00149947 InChI Key: KHCPSOMSJYAQSY-UHFFFAOYSA-L Synonym: platinum 4+ ion hydrate hydrochloride tetraazanide IUPAC Name: platinum(2+) tetraamine dichloride SMILES: N.N.N.N.[Cl-].[Cl-].[Pt++]