CAS RN 106372-55-8
CAS RN 106372-55-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Acesulfame Aspartame Salt, TRC
CAS: 106372-55-8 Synonym: Acesulfame Aspartame Salt∣Aspartame Acesulfame