CAS RN 1049747-87-6
CAS RN 1049747-87-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SB 242084 dihydrochloride, MedChemExpress
MedChemExpress SB 242084 hydrochloride is a 5-HT2C receptor antagonist(pKi=9.0) that displays 158- and 100-fold selectivity over 5-HT2A and 5-HT2B receptors respectively.