CAS RN 104968-58-3
CAS RN 104968-58-3
IUPAC Name:
1-isothiocyanato-3,5-dimethoxybenzene
Synonyms:
1-isothiocyanato-3,5-dimethoxybenzene
Molecular Weight (g/mol):
195.24
Molecular Formula:
C9H9NO2S
InChi Key:
FKUHOOASBHTEQY-UHFFFAOYSA-N
SMILES:
COC1=CC(=CC(OC)=C1)N=C=S
1
–
3
of
3
results
1
3,5-Dimethoxyphenyl Isothiocyanate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,5-Dimethoxyphenyl Isothiocyanate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,5-Dimethoxyphenyl Isothiocyanate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.