accessibility menu, dialog, popup

UserName

CAS RN 103909-75-7

CH3(S)OH3CH3COH(S)(S)(S)CH3(R)OH(S)HOH2C(E)(Z)

CAS RN 103909-75-7

IUPAC Name: (1R,3S,5Z)-5-{2-[(1S,3aS,4E,7aS)-1-[(1S)-1-(3-hydroxy-3-methylbutoxy)ethyl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexane-1,3-diol
Synonyms: maxacalcitol OCT oxarol prezios 22-oxa-calcitriol 马沙骨化醇 maxacalcitriol 22-oxacalcitriol
Molecular Weight (g/mol): 418.62
Molecular Formula: C26H42O4
InChi Key: DTXXSJZBSTYZKE-ZDQKKZTESA-N
SMILES: C[C@H](OCCC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)C[C@H](O)C1=C
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)