CAS RN 103336-06-7
CAS RN 103336-06-7
IUPAC Name:
1-[(tert-butoxy)carbonyl]-2-methylpyrrolidine-2-carboxylic acid
Synonyms:
1-(tert-butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid
Molecular Weight (g/mol):
229.28
Molecular Formula:
C11H19NO4
InChi Key:
YQXRKJHVAUKXRN-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)N1CCCC1(C)C(O)=O
1
–
1
of
1
results
1
N-Boc-2-methyl-L-proline, 97%, Thermo Scientific™
CAS: 103336-06-7 Molecular Formula: C11H19NO4 Molecular Weight (g/mol): 229.28 MDL Number: MFCD08706361 InChI Key: YQXRKJHVAUKXRN-UHFFFAOYNA-N Synonym: s-1-tert-butoxycarbonyl-2-methylpyrrolidine-2-carboxylic acid,1-tert-butoxycarbonyl-2-methyl-l-proline,1-boc-2-methyl-l-proline,n-boc-2-methyl-l-proline,n-boc-alpha-methyl-l-proline,2s-1-tert-butoxy carbonyl-2-methylpyrrolidine-2-carboxylic acid,2s-1-tert-butoxycarbonyl-2-methylpyrrolidine-2-carboxylic acid,n-boc-,a-methyl-l-proline,boc-alpha-me-l-pro-oh,s-1-boc-2-methyl-proline PubChem CID: 11206950 SMILES: CC(C)(C)OC(=O)N1CCCC1(C)C(O)=O