accessibility menu, dialog, popup

UserName

CAS RN 10190-55-3

PbOMoOOO

CAS RN 10190-55-3

IUPAC Name: λ²-lead(2+) dioxomolybdenumbis(olate)
Synonyms: λ²-lead(2+) molybdate
Molecular Weight (g/mol): 367.1
Molecular Formula: MoO4Pb
InChi Key: XJUNRGGMKUAPAP-UHFFFAOYSA-N
SMILES: [Pb++].[O-][Mo]([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results