accessibility menu, dialog, popup

UserName

CAS RN 1018134-12-7

CH3NCH3OCl

CAS RN 1018134-12-7

IUPAC Name: 2-chloro-N-ethyl-N-(3-methylphenyl)benzamide
Synonyms: 2-chloro-N-ethyl-N-(3-methylphenyl)benzamide
Molecular Weight (g/mol): 273.76
Molecular Formula: C16H16ClNO
InChi Key: YFBNVKAYRHEGFA-UHFFFAOYSA-N
SMILES: CCN(C(=O)C1=CC=CC=C1Cl)C1=CC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results