Learn More
Zinc Sulfate Solution 0.05M (0.05N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 Ampule
6 Ampules
Specifications
Melting Point | 0°C |
Boiling Point | 100°C |
Quantity | 1 Ampule |
Packaging | Ampoule |
Form | Liquid |