missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Heptahydrate, 99.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals

Catalog Number p-3547391
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
7446-20-0
H14O11SZn
287.54
MFCD00149894
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
62640
CHEBI:32312
zinc(2+) heptahydrate sulfate
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7446-20-0
H14O11SZn
287.54
MFCD00149894
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
62640
CHEBI:32312
zinc(2+) heptahydrate sulfate
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.0-102.0%
Formula Weight 287.54
Grade ACS Reagent
Chemical Name or Material Zinc Sulfate Heptahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'