Learn More
Masterflex™ Fitting, PVDF, Y-Connector, Hose Barb Union,
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
Brand:  Masterflex™ 30703-94
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11726289
  • 161.00 EUR / Pack of 10
Estimated Shipment
12-04-2021
Add to basket
Description
Description
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
Specifications
Specifications
Translucent
Yes
2 1/2 in.
Flexible Tubing
10/pk.
3
Barbed Y-Connector, PVDF, 1/2 in. ID
PVDF
I/P 82 B/T 88
Low pressure applications in pharmaceuticals, laboratory, industrial/manufacturing, water sampling or food processing
12.7 mm
Autoclave, Ethylene Oxide