Learn More
RETSCH XS4002S EasySieve Software Balance
Avoid random errors and operator errors in the evaluation of sieve analysis. Retsch™ XS4002S EasySieve Software Balance is controlled through RS232 serial interface for easy evaluation and documentation of results.
Brand: RETSCH 32.900.0001
Additional Details : Weight : 6.90000kg
Description
- For use with Retsch sieve shakers
- Validated according to DIN ISO IEC 12 119
- Takes over operator guidance and provides instructions about the next working step to be carried out throughout the whole sieving process
- Reduces the total time required for a sieve analysis by approximately 60 to 70% compared with a manual method
- Communicates with the balance and sieve shaker and calculates the sieving result as a cumulative distribution and frequency density
- Permits the calculation of the relevant parameters
- Graphic and tabular presentations of the sieving result are shown as well as the general sieving data in a well laid out standardized measuring protocol
- Trend analyses, comparative presentations, mean value calculations and data export are possible
Specifications
EasySieve Software Balance XS4002S | |
200/203 mm | |
EasySieve Evaluation Software |