Learn More
Lonza X-VIVO™ 15
Optimized for the proliferation of Tumor Infiltrating Lymphocytes (TIL) under serum-free conditions. X-VIVO™ 15 is a chemically defined, serum-free hematopoietic cell medium.
Brand: Lonza BE02-060Q
Additional Details : Weight : 0.60000kg
Description
Formulated with:- L-glutamine
- gentamicin
- phenol red
Order Info
dimProductType
Specifications
Cell Culture Media |