Learn More
WTW™ WTW™ Rectangular Cuvette
For automatic detection of the photLab™ filter and spectral photometer
77.00 EUR - 146.00 EUR
Product Code | Brand | Path Length | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Path Length | Price | Quantity & Availability | |||||
10508381
|
WTW™
250614 |
50 mm |
142.00 EUR
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
10324781
|
WTW™
250600 |
20 mm |
146.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10693372
|
WTW™
250605 |
10 mm |
77.00 EUR
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer. When using disposable cuvettes, the material PMMA as well as a structured or matte side surface is required due to the cuvette detection by the photoLab™ photometers. The glass and quartz cuvettes have a marking in order to place the cuvettes into the photometer in the same direction for an even higher precision.Cuvettes for turbidity measurement can be marked to prevent influences by glass: This allows a higher accuracy, also for values < 1 NTU.Specifications
Cuvette | |
Rectangle |