Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Aldrich FT-NMR Library with Spectral Viewer on CD-ROM
Allows for Spectra processing using Zoom, Peak Integration and Peak Labeling. The Bel-Art SP Scienceware™ Wilmad™ Aldrich FT-NMR Library with Spectral Viewer on CD-ROM is an electronic book that contains high-resolution 300MHz 1H and 75MHz 13C spectra, integral, chemical structure and technical data for the more than 11,800 compounds.
815.00 EUR - 1780.00 EUR
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15338698
|
Bel-Art™
66000-0107 |
1,1800 Compound Database |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15328698
|
Bel-Art™
66000-0106 |
3,500 Compound Database |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Supplemental library that includes spectra for 3,500 additional compounds not included in the original printed version is also available
- Order both the standard and the supplemental library for access to spectra on over 15,000 compounds
- Multiple search methods: text Search by Chemical Name, Aldrich Catalog Number, CAS Number, Molecular Formula, advanced data field search on chemical categories and physical properties, such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight and Refractive Index
- Minimum System Requirement: Windows Platform, CD-ROM drive, 600MB hard disk space
Specifications
FT-NMR Library with Spectral Viewer on CD-ROM |