Learn More
Motic™ Widefield Eyepiece WF5X/22mm
Widen your field of view for more complete specimen viewing. Motic™ Widefield Eyepiece WF5X/22mm provides an improved field of view and greater eye relief.
Brand:  Motic™ 51020141
Product Code. 11868183
  • 45.10 EUR / Each
Only null left
Estimated Shipment: 11-09-2025
Description
  • Widefield style provides improved field of view and greater eye relief.
  • Magnification: 5X.
  • Field of view: 22mm.
Specifications
Specifications
Widefield
FOV: 22mm
ST-39, SFC-11, SMZ-140 Microscopes
5X