Learn More
Motic™ Widefield Eyepiece WF30X/8mm
Widen your field of view for more complete specimen viewing. Motic™ Widefield Eyepiece WF30X/8mm provides an improved field of view and greater eye relief.
Brand: Motic™ SG02T0702
Additional Details : Weight : 0.80000kg
Description
- Widefield style provides improved field of view and greater eye relief.
- Magnification: 30X.
- Field of view: 8mm.
Specifications
Widefield | |
FOV: 8mm | |
DM-39, DM-143, ST-39 Microscopes | |
30X |