Learn More
Motic™ Widefield Eyepiece WF30X/8mm
Widen your field of view for more complete specimen viewing. Motic™ Widefield Eyepiece WF30X/8mm provides an improved field of view and greater eye relief.
Brand:  Motic™ SG02T0702
Additional Details : Weight : 0.80000kg
Product Code. 11821930
  • 74.00 EUR / Each
Estimated Shipment: 29-07-2024
Description
  • Widefield style provides improved field of view and greater eye relief.
  • Magnification: 30X.
  • Field of view: 8mm.
Specifications
Specifications
Widefield
FOV: 8mm
DM-39, DM-143, ST-39 Microscopes
30X