missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Reagent Bottle, Wide Neck, Clear, Without Stopper
283.10 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
250 mL
50 mL
500 mL
5000 mL
Description
The classical stoppered reagent bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a wide conical ground joint neck and comply with ISO 4796-2. The wider neck is more convenient for handling powders. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel.
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- All borosilicate glass components and therefore suitable for storage of aggressive materials
- Tight closure due to standard ground joint
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
Specifications
Diameter (Metric) | 44 mm |
Capacity (Metric) | 50 mL |
Description | Neck with standard ground joint |
For Use With (Application) | Storage of air sensitive reagents, pharmaceuticals, and chemicals; Storage of hygroscopic liquids and pharmaceuticals; Transport and shipping of samples |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'