Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners
Brand:  DWK Life Sciences 239216
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11778829
  • 35.41 EUR / Pack of 72
Estimated Shipment
19-03-2021
Add to basket
Description
Description
  • White polypropylene screw cap provides clear surface for writing on cap
  • Poly-vinyl liner provides an excellent moisture barrier
Specifications
Specifications
ISO 9001
Round bottle
Polypropylene
Polypropylene Cap
Poly/Vinyl
White
72 Cs.
48-400