Learn More
DWK Life Sciences Wheaton™ Vial File™ with Screw Cap: Amber
Amber sample vials with a variety of screw cap options
110.00 EUR - 237.00 EUR
Specifications
Color | Amber |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Used for storing and transporting vials |
Thread Style | Screw |
Closure Material | Phenolic |
Product Code | Brand | Closure Type | Diameter (Metric) Outer | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Type | Diameter (Metric) Outer | Volume (Metric) | Price | Quantity & Availability | |||
12424801
|
DWK Life Sciences
W224954 |
Screw Cap (Open-top) | 12 mm | 1.5 mL |
134.00 EUR
Pack of 60 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
12414801
|
DWK Life Sciences
W224897 |
Screw Cap | 15 mm | 4 mL |
237.00 EUR
Pack of 40 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
12434801
|
DWK Life Sciences
W224955 |
Screw Cap (Open-top) | 15 mm | 4 mL |
110.00 EUR
Pack of 40 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
Description
Amber sample vials with a variety of screw cap options
The Vial File provides a compact, easily accessible means of sample storage. The vials are stored in individual partitions within a clear polystyrene case. Foam inserts provide extra protection for the vials.
- Alpha numerical indexing card allows for identification of each sample vial
- Vials are manufactured from borosilicate glass which provides superior chemical resistance
- Cat. Nos. 02-912-591 and -597 feature open-top black phenolic screw cap for easy access with a syringe
- The Teflon-faced silicone lined screw cap is ideal for low-temperature applications; provides excellent chemical barrier
- Vials and caps are autoclavable
Specifications
Amber | |
Used for storing and transporting vials | |
Phenolic |
Borosilicate Glass | |
Screw | |
USP Type I |