Learn More
DWK Life Sciences Wheaton™ Solid Top Polypropylene Screw Caps
26.84 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Use promo code "23261" to get your promotional price.
Click to view available options
PP; White; w/ Poly-Vinyl liner; Non-autoclavable; 144/Cs.
Description
- Used with glass bottles or sample vials
- Non-autoclavable
- White colored
Specifications
Closure Material | Polypropylene |
Color | White |
Material | Polypropylene |
Quantity | 144/Cs. |
Thread Size (Metric) | 13-425 |
Liner Material | Polyvinyl |
Autoclavable | Not Autoclavable |