Learn More
DWK Life Sciences Wheaton™ Septa for Open-Top Caps
Can be used with screw caps
Brand:  DWK Life Sciences W240596
120.65 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 10297123
  • 127.00 EUR / Pack of 100
Estimated Shipment: 01-10-2024
Description
  • For caps with open tops
  • Autoclavable
Specifications
Specifications
PTFE, Rubber
PTFE-faced natural rubber
Septa
Sterile
Autoclavable
20 mm