Learn More
DWK Life Sciences Wheaton™ Septa For Unlined Aluminum Seals
Click to view available options
Diameter (English):
0.5 in.
0.7 in.
1.1 in.
Color:
Gray
Red
Material:
PTFE-faced Butyl
PTFE-faced Silicone
Silicone
Specifications
Closure Size | 20 mm |
Diameter (Metric) | 20 mm |
For Use With (Equipment) | For unlined aluminum seals |
Color | Red |
Diameter (English) | 0.7 in. |
Material | Silicone |
Quantity | 1 Cs. |