Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Gray
Product Code. 12406161 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
13-425
15-425
20-410
8-425
No. per Case:
100/Cs.
1000/Cs.
Unit Size:
Pack of 100
Pack of 1000
Product Code. 12406161
Brand: DWK Life Sciences W242535A
Only null left
Add to basket
Add to basket
Screw cap made of radiation grade polypropylene
  • Packaged in case of 100 or 1000
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Material Polypropylene
No. per Case 1000/Cs.
Neck Finish 13-425
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Plastic dropping bottle