Learn More
DWK Life Sciences Wheaton™ Jars with Polypropylene Screw Cap
Product Code. p-4380944 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Autoclavable:
Autoclavable (Bottle Only)
Not Autoclavable
Color:
Clear
Natural
White
Unit Size:
Pack of 24
Pack of 36
Pack of 48
Pack of 72
Product Code. 12437391
Brand: DWK Life Sciences W209912
Only null left
Add to basket
Add to basket
  • Good resistance to inorganic chemicals
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner
TRUSTED_SUSTAINABILITY
Specifications
Capacity 1.01 oz., 30mL
Capacity (Metric) 30 mL
Capacity (English) 1.01 oz.
Color Clear
Diameter (English) 1.53 in.
Includes Screw Cap
Material Polystyrene
Closure Color White
Clarity Clear
Mouth Wide
Shape Round
Height (Metric) 41 mm
Product Type Jar
Height (English) 1.61 in.
Closure Liner Polyethylene Foam
Closure Material Polypropylene
Closure Size 43-400 mm
Diameter (Metric) 39 mm
Closure Type Screw Cap