Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Replacement for Double Sidearm Celstir and Jacketed Double Sidearm Celstir.
Brand:  DWK Life Sciences 356895
178.60 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 12421421
  • 188.00 EUR / Each
Estimated Shipment: 23-10-2024
Description
  • This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
Specifications
Specifications
50mL Celstir™ Spinner Flask
Top cap with liners, glass rod, magnet holder, magnet and stirring paddle