Learn More
DWK Life Sciences Wheaton™ E-Z Seals™ Aluminum Vial Caps
Lined seals for use with crimp-top vials
Brand: DWK Life Sciences 224219-01
454.10 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.45360kg
Description
- Ideal closures for chromatography and other instrumentation applications
- Open top seal provides for filling or retrieving contents with a syringe
Specifications
PTFE/Silicone | |
Aluminum | |
Open Top Cap |
Natural | |
100 Pk. | |
11 mm |