Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with PE Cone-Lined Caps
Ideal for solvent, chemical and sample storage
29.05 EUR - 79.60 EUR
Specifications
| Color | Clear |
|---|---|
| Material | Soda Lime Glass |
| Closure Color | Black |
| Certifications/Compliance | EPA, FDA |
| Closure Liner | Polyethylene |
| Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|
12379729
|
DWK Life Sciences
W216812 |
30 mL | 20-400 mm |
79.60 EUR
Pack of 48 |
Estimated Shipment: 08-04-2026
Log in to see stock. |
||||
|
12389729
|
DWK Life Sciences
W216813 |
60 mL | 20-400 mm |
33.65 EUR
Pack of 24 |
Estimated Shipment: 21-11-2025
Log in to see stock. |
||||
|
12399729
|
DWK Life Sciences
W216814 |
125 mL | 22-400 mm |
42.35 EUR
Pack of 24 |
Estimated Shipment: 08-04-2026
Log in to see stock. |
||||
|
12309739
|
DWK Life Sciences
W216815 |
250 mL | 24-400 mm |
29.05 EUR
Pack of 12 |
Estimated Shipment: 08-04-2026
Log in to see stock. |
||||
|
12319739
|
DWK Life Sciences
W216816 |
500 mL | 28-400 mm |
33.20 EUR
Pack of 12 |
Estimated Shipment: 08-04-2026
Log in to see stock. |
||||
|
12329739
|
DWK Life Sciences
W216817 |
1000 mL | 33-400 mm |
46.70 EUR
Pack of 12 |
Estimated Shipment: 08-04-2026
Log in to see stock. |
||||
Description
- Well suited for storage and sampling of liquids; clear Type III soda-lime glass provides visibility of contents
- Narrow mouth enables liquids to be carefully poured to prevent spillage
Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage.
Order Info
Caps can be ordered separately.
Specifications
| Clear | |
| Black | |
| Polyethylene | |
| Screw Cap |
| Soda Lime Glass | |
| EPA, FDA | |
| Phenolic |