Learn More
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Brand:  DWK Life Sciences 224187-01
155.80 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 1.08860kg
 View more versions of this product
Product Code. 10331772
  • 164.00 EUR / Pack of 1000
Estimated Shipment: 30-05-2024
Description
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among others
Specifications
Specifications
Natural
1000/Cs.
30 mm
Aluminum
Center Disc Tear-out