Learn More
Cytiva Whatman™ Mini-Uniprep™ Syringeless Filter for HPLC Robotics
Provide a faster filtering particulates from HPLC and UHPLC samples providing 67% time and 40% monetary savings
Brand:  Cytiva US503NPUGMF
Additional Details : Weight : 3.07000kg
Product Code. 11328784
  • 2515.00 EUR / Pack of 1000
Estimated Shipment: 19-07-2024
Description
Slit septum cap enables Mini-UniPrep use with current robotics on HPLC instruments for high throughput automation. These durable yet flexible slit septum caps have been specially designed for instruments with sensitive sampling needs. Sample evaporation is minimal.
  • GMF 150 Glass Microfiber Filter; Retention (Pore Size): 0.45μm
  • Slit septum cap enables Mini-UniPrep™ use with standard robotics on HPLC instruments with sensitive needles, allowing for higher throughput
  • Processing samples through the syringeless filter can be performed either manually with the furnished plunger or with a compressor unit
  • Designed to fit into any autosampler accommodating 12×32mm vials
  • Pack of 1000
Specifications
Specifications
0.45 μm