Description
- New material MICROMIUM™, light and very low contaminant
- Cleanroom apparel compatible from Iso class 4
- Standardised chemical splash suit, Category 3 Type 5/6
- Double packaging fully sealed
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Material | Microporous Polyethylene Laminate |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic (Stitched-in) |
| Ankle Style | Covered Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |