Learn More
Medicom WeePro Labo Coverall, Type 5/6
GREENER_CHOICE
Product Code. 17879291 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 25
Product Code. Size unitSize
17879291 X-Large Pack of 25
17859301 Medium Pack of 25
17869301 Large Pack of 25
17879301 2X-Large Pack of 25
4 options
Product Code. 17879291
Brand: Medicom WLPLM04
Standardized chemical splash suit that is cleanroom apparel compatible from ISO class 4.
  • New material MICROMIUM™, light and very low contaminant
  • Cleanroom apparel compatible from Iso class 4
  • Standardised chemical splash suit, Category 3 Type 5/6
  • Double packaging fully sealed
Specifications
Product Type Cleanroom Coveralls
Size X-Large
Gender Unisex
Material Microporous Polyethylene Laminate
Color White
Disposable Single-use
Waist Style Elastic (Stitched-in)
Ankle Style Covered Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type 2-way Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals
Latex Free No
No. of Pockets None
Cleanroom Class Class 10-100,000 (ISO 4-8)