Learn More
Emma Safety Footwear Vulcanus Safety Shoes
Welder's boot designed specifically for working with metal in extreme temperatures. Over-foot flap provides no entry for sparks or shards.
Brand: Emma Safety Footwear MM53984841
Additional Details : Weight : 1.89900kg
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Velcro over-foot flap
- Sanitized
Specifications
EN ISO 20345:2011 | |
Vulcanus | |
Leather |
Black | |
Full grain Nappa /Hydro-Tec™ Sanitized Silver | |
41 |