Learn More
Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask with PP Stopper Conformity Certified is thick-walled, clearly marked and labeled, and has a solid finish
Brand:  Karl Hecht™ 42096008
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10206961
  • 14.05 EUR / Each
Estimated Shipment: 03-07-2024
Description
  • Duran™ borosilicate glass 3.3.
  • Standard type.
  • Standard ground and interchangeable NS stopper of polyethylene.
  • Fused blue graduation.
  • Thick-walled, solid finish.
  • Clearly legible figures and marks.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Specifications
DIN EN ISO 1042
5 mL
Wide
0.04 mL
NS Stopper
Polyethylene
Borosilicate Glass (3.3)
Volumetric Flask
Yes
NS 10/19