Learn More
Poulten Graf™ Volac™ USP Class A Soda Lime Glass Bulb Pipette with Works Certificate
Individually marked with the time of flow to transfer. Poulten Graf™ Volac™ USP Class A Soda Lime Glass Bulb Pipettes with Works Certificate are guaranteed to give both volumes and flow times within Class A limits.
Brand:  Poulten Graf™ US371/WAC/B/5
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10000422
  • 319.00 EUR / Pack of 5
Estimated Shipment
14-03-2019
Add to basket
Description
Description
  • Individually numbered and meets the accuracies of USP/ISO/BS and DIN standards
  • Amber stain graduation
  • Calibrated for delivery
Specifications
Specifications
Certificate of calibration/Satisfying USP/ISO/BS and DIN accuracies
Yes
0.006mL
2mL
A
5Pack
Bulb Pipette
Soda Lime Glass
Videos