Learn More
Sartorius™ Vivaflow™ 50 Complete System
Product Code. 10342312 Shop All Sartorius Products
Click to view available options
Volume (Metric):
100 to 3000 mL
Unit Size:
Each
Product Code. 10342312
Brand: Sartorius™ VFS502
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material Polycarbonate
Material (Membrane) TPX
Filtration Area 50 cm2
For Use With (Application) Optimal for aqueous sample concentration e.g. proteins and nanoparticles
Housing Material Polycarbonate
Max. Pressure 3bar
Pump Flow Rate 200 to 400mL/min.
Quantity 1 Each
Recirculation Rate 10mL
Temperature (Metric) Max 60°C
Type Vivaflow 50 Complete System
Volume (Metric) 100 to 3000 mL
Volume (Metric) Hold Up 1.5 mL
Width (Metric) 25 mm
Width (Metric) Channel 15 mm
Height (Metric) 84 mm
Height (Metric) Channel 0.3 mm