missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Vivaflow 50

Product Code. 15700538 Shop All Sartorius Products
Click to view available options
:
100 to 3,000 mL
This item is not returnable. View return policy

Product Code. 15700538

Brand: Sartorius Stedim Biotech™ VF05P6

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Disposable and ready-to-use crossflow cassette for filtration and concentration of samples from 100 mL to 3L.

  • Plug and play laboratory cross flow cassette
  • Modular - scalable t0 6 units
  • Optimal for aqueous sample concentration, e.g., proteins and nanoparticles
  • High flow rate and recover
  • Clear housing for process monitoring
  • Easy set up
  • Economical and disposable
TRUSTED_SUSTAINABILITY

Specifications

Material PVC, Silicone, Polyethersulfone, Polypropylene, Nylon
MWCO 1000 kDa
Filtration Area 50 cm2
Max. Pressure 3 bar
Pump Flow Rate 200 to 400 mL/min.
Sample Type Nanoparticles
Temperature (Metric) Max 60°C
Type Tangential/Crossflow Concentrator
Volume (Metric) 100 to 3000 mL
Width (Metric) 25 mm
Width (Metric) Channel 15 mm
Height (Metric) 84 mm
Height (Metric) Channel 0.3 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.