Learn More
Macherey-Nagel™ Visocolor™ ECO Test Kit
Allows determination of even low limiting values with accuracy. Macherey-Nagel™ Visocolor™ ECO Test Kits is a group of colorimetric and titrimetric test kits that use precisely dosed single reagents. The results are evaluated visually with high-quality color comparison cards.
53.90 EUR - 101.00 EUR
Specifications
Includes | Reagents, Color comparator, Measuring spoon/glass, Syringe, Instructions in a cardboard box with plastic insert |
---|---|
Quantity | 1 Each |
Product Code | Brand | For Use With (Application) | Test Method | No. of Tests | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Test Method | No. of Tests | Price | Quantity & Availability | |||
10457292
|
Macherey-Nagel™
931040 |
Nickel determination | Colorimetric | 150 Determinations |
91.00 EUR
1 test |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||
10378551
|
Macherey-Nagel™
931023 |
Cyanuric acid determination | Colorimetric | 100 Determinations |
101.00 EUR
1 test |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||
10570403
|
Macherey-Nagel™
931024 |
DEHA determination | Colorimetric | 125 Determinations |
99.60 EUR
1 test |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
10054022
|
Macherey-Nagel™
931016 |
Free Chlorine determination | Colorimetric | 150 Determinations |
53.90 EUR
150 tests |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||
Description
Visocolor™ ECO colorimetric and titrimetric test kits achieve high sensitivity and accuracy by using single reagents that can be dosed precisely and by the ability to compensate for the turbidity and color of water samples. The results are evaluated visually with color comparison cards adjusted to the original colors of standard solutions or photometrically with the PF-3 and PF-12Plus photometers for quantitative evaluation.
Easy
- Chemical analysis without further accessories
- No extensive training necessary
- Color-coded reagents with clear dosing instructions
- Easy to understand instruction manual
Safe
- Pictogram test instructions
- Reaction basis according to international standards
- Compensation of turbidity and color
Unique
- High quality test kits
- Business-prized refill packs available
- Ecologically friendly disposal of used reagents
- Delivered in cardboard box with plastic inlay
Specifications
Reagents, Color comparator, Measuring spoon/glass, Syringe, Instructions in a cardboard box with plastic insert | |
1 Each |