missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ark Plas™ Vinyl (PVC) Tubing
For flexible, easily routed plastic tubing
Brand: Ark Plas™ TCX22-FC1
Additional Details : Weight : 0.27000kg
Description
Specifications
Black | |
1.5875 mm | |
3.175 mm | |
0.79375 mm |
Durometer: 90 Shore A | |
PVC | |
30 m |