Learn More
Brady™ Vinyl Coated Metal Cable
Sheathed Metal Cable for use with All Purpose Cable Lockout.
Brand: Brady™ 813642
Additional Details : Weight : 0.12800kg
Beschreibung
- Sheathed metal cable for use with All Purpose Cable Lockout.
Spezifikation
Lockout Cable | |
Vinyl Coated Metal Cable - 3 m | |
4 mm | |
3.05 m |
Red | |
0.16 in. | |
0.12 in. | |
1 Ea. |